NU1025
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX270370M005 ALX270370M025
ENZOALX270370M005EA 173 GBP
ENZOALX270370M005 ENZOALX270370M025
NU1025
NU1025
PARP-1 inhibitor
A potent poly(ADP-ribose) polymerase 1 (PARP-1) inhibitor (IC50=400nM) that potentiates the cytotoxicity of various DNA-active agents, including the DNA-methylating compound MTIC, the DNA strand break-inducing drug temozolomide (Prod. No. ALX-420-044), topotecan (Prod. No. ALX-350-133), bleomycin (Prod. No. BML-AP302), and ionizing radiation effective in murine L1210 leukemia cells, CHO, and in a variety of human tumor cell lines.
CAS Number: 90417-38-2