Z-DEVD-FMK
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX260141R020 ALX260141R100
ENZOALX260141R020EA 282 GBP
ENZOALX260141R020 ENZOALX260141R100
Z-DEVD-FMK
Proteins and Peptides
Inhibitor of caspase-3 and -7
Irreversible, cell permeable inhibitor of caspase-3 and -7.