Z-WEHD-FMK
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX260140R020 ALX260140R100
ENZOALX260140R020EA 276 GBP
ENZOALX260140R020 ENZOALX260140R100
Z-WEHD-FMK
Proteins and Peptides
Inhibitor of caspase-1, -4 and -5
Irreversible, cell permeable inhibitor of caspase-1 and -5. Inhibits also caspase-4. Designed as a methyl ester to facilitate cell permeability.