Actinonin ≥98%
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX260128M005
ENZOALX260128M005EA 173 GBP
ENZOALX260128M005
Actinonin ≥98%
Actinonin
Protease inhibitor
Antibiotic. Inhibitor of aminopeptidase M, N and leucine and alanyl aminopeptidase and peptide deformylase (PDF). Inhibits MMP-1 (Ki=300nM), MMP-2 (IC50~90nM at MMP-2=5nM), MMP-3 (Ki=1’700nM), MMP-7 (IC50~3’500nM at MMP-7=19nM), MMP-8 (Ki=160nM), MMP-9 (Ki=330nM), MMP-10 (IC50~500nM at MMP-10=22nM), MMP-12 (IC50~350nM at MMP-12=4nM), and MMP-13 (IC50~100nM  at MMP-13=13nM), but not MMP-14 (IC50>1mM at MMP-14=13.6nM). Shows antitumor activity in vitro and in vivo. Apoptosis inducer.
Synthetic.
CAS Number: 13434-13-4