Z-YVAD(OMe)-FMK
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX260074M001
ENZOALX260074M001EA 570 GBP
ENZOALX260074M001
Z-YVAD(OMe)-FMK
Proteins and Peptides
Inhibitor of caspase-1 and -4
Irreversible, cell permeable inhibitor of caspase-1. Inhibits also caspase-4.