Anti-MTOR Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX2150651
ENZOALX2150651EA 665 GBP
ENZOALX2150651
Anti-MTOR Rabbit Polyclonal Antibody
Antibodies
mTOR (FKBP12-rapamycin associated protein (FRAP)) is one of a family of proteins involved in cell cycle progression, DNA recombination, and DNA damage detection. It has been shown to associate with the immunophilin FKBP12 in a rapamycin-dependent fashion. FKBP12-rapamycin-binding (FRB) domain is a conserved 11kDa region necessary for FKBP12-rapamycin binding. FRB is one of several functional domains of mTOR: HEAT (Huntington elongation factor 1A-protein phosphatase 2A-A subunit-TOR); FAT (FRAP, ATM, TTRAP2); PIKK (PI3-kinase-related kinase); RD (regulatory domain); and FATC (FAT, C terminal).More Product Information
Order Now

SPECIFICATIONS

Antigen symbol MTOR
Antigen name Mechanistic Target of Rapamycin
Environmentally Preferable
Antigen synonyms FRAP1|mTOR|m-TOR|RAPT1|RAFT1|FKBP-Rapamycin Associated Protein|Mechanistic Target Of Rapamycin|EC 2.7.11.1|FRAP|FRAP2|Rapamycin Target Protein|FK506 binding protein 12-rapamycin associated protein|Mammalian Target Of Rapamycin|Serine/Threonine-Protein Kinase MTOR|MTOR
Antibody type Primary
Clonality Polyclonal
Reactivity Multiple
Host Rabbit
Isotype
Western blot Yes
ImmunoChemistry Yes
ChIP Yes
Form Lyophilized from neat serum containing sodium azide.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR