Anti-MDA5 Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ALX210935C100
ENZOALX210935C100EA 534 GBP
ENZOALX210935C100
Anti-MDA5 Rabbit Polyclonal Antibody
Antibodies
MDA5 is a highly conserved helicase whose ATPase activity can be stimulated by RNA and less potently by DNA. It contains two N-terminal CARD domains and a C-terminal helicase domain. The helicase domain, once freed from the CARD domains, then translocates from the cytoplasm to the nucleus where it acts on chromatin architecture. This could allow a more easy access of CAD, thereby accelerating the degradation of genomic DNA.

  • For Immunoprecipitation, Western Blot

Recognizes human MDA5.

Type: Primary
Antigen: MDA5
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR