Human recombinant Matriptase
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ALX201246U250
ENZOALX201246U250EA 671 GBP
ENZOALX201246U250
Human recombinant Matriptase
Enzymes
Recombinant human matriptase (aa 596-855) fused at the N-terminus to a His-tag.