Anti-HSP91 Rat Monoclonal Antibody
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADISPA845D
ENZOADISPA845DEA 261 GBP
ENZOADISPA845D
Anti-HSP91 Rat Monoclonal Antibody
Antibodies
The 90kDa molecular chaperone family includes 90 kDa heat shock protein Hsp90 and 94 kDa glucose-regulated protein grp94, both major molecular chaperones of the cytosol and the endoplasmic reticulum. Mammalian cells contain isoforms Hsp90α and Hsp90β, encoded by separate genes. The amino acid sequences of human and yeast Hsp90-alpha are 85% and 90% homologous to those of Hsp90β , respectively. All known members of the Hsp90 protein family are highly conserved, especially in the N-terminal and C-terminal regions containing independent chaperone sites with different substrate specificity.More Product Information