Anti-EC SOD Rabbit Polyclonal Antibody

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ADISOD106D ADISOD106F
ENZOADISOD106DEA 242 GBP
ENZOADISOD106D ENZOADISOD106F
Anti-EC SOD Rabbit Polyclonal Antibody
Antibodies
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).

Type: Primary
Antigen: EC SOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR