Brain (rat) tissue extract

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ADILYTRB100F
ENZOADILYTRB100FEA 256 GBP
ENZOADILYTRB100F
Brain (rat) tissue extract
Specimens Preserved Specimens Preserved Animal Materials
  • Liquid

Delivery information: SDS-PAGE sample buffer (pH: 6,8) containing 40 mM TRIS-HCl, 1% SDS, 50mM DTT, 7,5% glycerol and 0,003% bromophenol blue.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR