ImmunoSet® αB-Crystallin ELISA development set

Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

ADI960074
ENZOADI960074EA 731 GBP
ENZOADI960074
ImmunoSet® αB-Crystallin ELISA development set
Assays ELISAs

For the quantitative determination of αB-Crystallin in cell lysates of human, mouse, rat, and cow origin.


The ImmunoSet® αB-Crystallin EIA development set contains sufficient reagents for the development of 5×96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.


Ordering information: Contains Capture antibody, Standard, Detection antibody, SA-HRP.

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein αB-Crystallin
Primary antibody reactivity Human,Bovine,Rat,Mouse
Host
Format Uncoated
Assay duration Multiple steps
Sensitivity 0,59 ng/ml (range 1,25 - 40 ng/ml)
Detection method Colorimetric 450 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR