Anti-Bradykinin Receptor 1 Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADI905787100
ENZOADI905787100EA 483 GBP
ENZOADI905787100
Anti-Bradykinin Receptor 1 Rabbit Polyclonal Antibody
Antibodies
Bradykinin (BK) is a nine-amino acid peptide produced by kallikrein cleavage of kininogen precursor proteins in tissue and plasma, and is an important regulator of vascular and pain responses to tissue injury. Two receptors (B1 and B2) have been identified as mediators of BK signaling, and are both members of the seven-transmembrane domain-containing G-protein coupled receptor (GPCR) family. The B2 receptor is constitutively expressed in smooth muscle and neurons, whereas B1 expression is induced following tissue injury or during inflammation. The B2 receptor displays high affinity for BKMore Product Information