cGMP ELISA kit
Supplier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADI901013
ENZOADI901013EA 2080 GBP
ENZOADI901013
cGMP ELISA kit
Assays ELISAs
Most sensitive (0,088 pmol/ml) colorimetric kit  for extracellular cGMP quantification.
For the quantitative determination of cGMP in culture supernatants, plasma, serum, saliva, and urine from any species.
This is a colorimetric competitive immunoassay kit for the quantitative determination of extracellular cGMP. Sensitivity is increased by acetylation (reagents included). Absorbance is read at 405 nm.The ready-to-use liquid color-coded reagents provided reduce error.
Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer 2 concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine. Alternative size available: 89141-066 (1×96 wells).