White light viewer, LabLite illuminator, ViewOne™
Supplier: EMBITEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VL-1000 VL-1200
EMBIVL-1000EA 217 GBP
EMBIVL-1000 EMBIVL-1200
White light viewer, LabLite illuminator, ViewOne™
Transilluminators LED Transilluminators
Compact benchtop illuminator helps to visually identify a specific sample well, complete lanes or rows, to aid in loading 96-well plates.
  • Uniform background for enhanced visualization of colorimetric samples
  • Waterproof top allows for visualizing wet samples such as SDS-PAGE gels or dissection samples
  • Portable and space saving
The ViewOne™ LabLite is a stand-alone bench top solution for visualizing colorimetric sample against a uniform white light background. Labs have used the ViewOne with ELISA plates, colony picking and bacterial plates,More Product Information