CpG methyltransferase

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

EM0821
EM0821EA 222 GBP
EM0821
CpG methyltransferase
Enzymes

CpG methyltransferase enzyme methylates the C5 position on the base moiety of all cytosine nucleotides contained in unmethylated or hemimethylated double stranded DNA in a 5'-CpG-3' context. Thermo Scientific M.SssI is specifically formulated for fast reaction times without compromising the reaction efficiency. The enzyme completes modification of all CpGs in 15 minutes at 37 °C. In addition, the enzyme has been specifically validated for use with genomic DNA – the primary substrate in epigenetic studies.


  • High efficiency: Complete in vitro methylation of all CpGs in non-methylated and hemi-methylated DNA
  • Fast: Reaction is completed in 15 minutes at 37 °C
  • Stable at −20 °C for up to 2 years


Delivery information: M.SssI is supplied with an optimised 10X reaction buffer and 50X S-adenosylmethionine (SAM) as a cofactor.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR