Cole-Parmer® SHM-200 Multiposition Extraction Heater/Stirrers
Supplier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03076-13 03076-16 03076-19 03076-22 03076-37 03076-40 03076-43
ELET03076-13EA 2670 GBP
ELET03076-13 ELET03076-16 ELET03076-19 ELET03076-22 ELET03076-37 ELET03076-40 ELET03076-43
Cole-Parmer® SHM-200 Multiposition Extraction Heater/Stirrers
Heaters Heating Mantles
Allows liquids with different boiling points to be heated and or stirred next to each other.
  • Stir or heat solutions simultaneously in each position
  • Accommodate flask capacities from 100 to 1000 ml
  • Unique vented airflow ensures the case remains 'cool-to-the-touch'
  • Robust construction permits continuous operation of mantles as required by routine extraction tests
  • Material: Aluminium
Regulating the heat on these multi-position extraction mantles is simple using the built-in solid-state electronic controls. The units accept round bottom flasks rangingMore Product Information