Cole-Parmer® HM-200 Multi-Position Extraction Heaters
Supplier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03076-01 03076-04 03076-07 03076-10 03076-25 03076-28 03076-31 03076-34
ELET03076-01EA 1410 GBP
ELET03076-01 ELET03076-04 ELET03076-07 ELET03076-10 ELET03076-25 ELET03076-28 ELET03076-31 ELET03076-34
Cole-Parmer® HM-200 Multi-Position Extraction Heaters
Heaters Heating Mantles
Allow liquids with different boiling points to be heated next to each other.
  • Vented airflow ensures the case remains 'cool to the touch'
  • Heat your solutions simultaneously in each position
  • Robust construction permits continuous operation of the mantles as required by routine extraction tests
Regulating the heat on these multi-position extraction mantles is simple using the built-in solid-state electronic controls. The units accept round-bottom flasks ranging from 100 to 1000 ml. Select an extraction heater depending on the size of your flask.