Pesticide reference standard, 2,3,5,6-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L17375600CY
EHERL17375600CYEA 135 GBP
EHERL17375600CY
Pesticide reference standard, 2,3,5,6-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Standards Organic Standards

Order Now

SPECIFICATIONS

Description 2,3,5,6-Tetrachlorophenol
Environmentally Preferable
CAS No. 935-95-5
Formula C₆H₂Cl₄O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR