2,4-Toluenediisocyanate

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CA17594500
EHERCA17594500EA 104 GBP
EHERCA17594500
2,4-Toluenediisocyanate
2,4-Toluenediisocyanate
Formula: C₉H₆N₂O₂
MDL Number: MFCD00002011
CAS Number: 584-84-9
EINECS: 209-544-5
UN: 2078
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR