2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: PCB No. 138, 2,2',3,4,4',5'-PCB

C20013800
EHERC20013800EA 194 GBP
EHERC20013800
2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)
2,2',3,4,4',5'-Hexachlorobiphenyl (PCB No. 138)
Formula: C₁₂H₄Cl₆
Storage Temperature: Refrigerator
CAS Number: 35065-28-2
UN: 2315
ADR: 9,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR