2,3',4'-Trichlorobiphenyl (PCB No. 33)

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C20003300
EHERC20003300EA 102 GBP
EHERC20003300
2,3',4'-Trichlorobiphenyl (PCB No. 33)
2,3',4'-Trichlorobiphenyl (PCB No. 33)
Formula: C₁₂H₇Cl₃
CAS Number: 38444-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR