(±)-2,2,4-Trimethyl-1,3-pentanediol

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C17883150
EHERC17883150EA 107 GBP
EHERC17883150
(±)-2,2,4-Trimethyl-1,3-pentanediol
(±)-2,2,4-Trimethyl-1,3-pentanediol
Formula: C₈H₁₈O₂
Boiling Pt: 228.9 °C (1013 hPa)
Melting Pt: 51 °C
Density: 0.9485 g/cm³ (20 °C)
MDL Number: MFCD00004681
CAS Number: 144-19-4
EINECS: 205-619-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR