Succinylsulphathiazole

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N4-Succinyl-N-(2-thiazolyl)sulphanilamide, 4-(Succinylamino)-N-(2-thiazolyl)benzenesulphonamide, 4-Oxo-4-((4-(N-(thiazol-2-yl)sulphamoyl)phenyl)amino)butanoic acid

C16985600
EHERC16985600EA 99.1 GBP
EHERC16985600
Succinylsulphathiazole
Succinylsulfathiazole
Formula: C₁₃H₁₃N₃O₅S₂
MW: 355.38 g/mol
MDL Number: MFCD00022437
CAS Number: 116-43-8
EINECS: 204-141-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR