Pyrazinamide

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide

C16608000
EHERC16608000EA 89.7 GBP
EHERC16608000
Pyrazinamide
Pyrazinamide
Formula: C₅H₅N₃O
MW: 123.11 g/mol
Boiling Pt: 357
Melting Pt: 188…191 °C
MDL Number: MFCD00006132
CAS Number: 98-96-4
EINECS: 202-717-6
Merck Index: 12,08140

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR