1-Propanol

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

C16415100
EHERC16415100EA 92 GBP
EHERC16415100
1-Propanol
1-Propanol
Formula: H₃CCH₂CH₂OH
Boiling Pt: 97 °C (1013 hPa)
Melting Pt: –127 °C
Density: 0.8053 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR