Procymidone

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C16310000
EHERC16310000EA 189 GBP
EHERC16310000
Procymidone
Procymidone
Formula: C₁₃H₁₁Cl₂NO₂
CAS Number: 32809-16-8
EINECS: 251-233-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR