Diphenyl phthalate

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

C16176000
EHERC16176000EA 104 GBP
EHERC16176000
Diphenyl phthalate
Diphenyl phthalate
Formula: C₂₀H₁₄O₄
Boiling Pt: 405 °C (1013 hPa)
Melting Pt: 74…76 °C
Density: 1.313 g/cm³ (20 °C)
MDL Number: MFCD00003038
CAS Number: 84-62-8
EINECS: 201-546-4
Merck Index: 12,07460

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR