Dihexyl phthalate

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C16173200
EHERC16173200EA 135 GBP
EHERC16173200
Dihexyl phthalate
Dihexyl phthalate
Formula: C₂₀H₃₀O₄
MDL Number: MFCD00043720
CAS Number: 84-75-3
EINECS: 201-559-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR