n-Pentadecane

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-Pentadecane, Pentadecane

C15973700
EHERC15973700EA 151 GBP
EHERC15973700
n-Pentadecane
n-Pentadecane
Formula: H₃C(CH₂)₁₃CH₃
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 13…15 °C
Density: 0.768 g/cm³ (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00008990
CAS Number: 629-62-9
EINECS: 211-098-1
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR