D-Panthenol

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Dexpanthenol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

C15844500
EHERC15844500EA 236 GBP
EHERC15844500
D-Panthenol
D-Panthenol
Formula: C₉H₁₉NO₄
MW: 205.25 g/mol
MDL Number: MFCD00065006
CAS Number: 81-13-0
EINECS: 201-327-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR