Orotic acid

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

C15747000
EHERC15747000EA 210 GBP
EHERC15747000
Orotic acid
Orotic acid
Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
MDL Number: MFCD00006027
CAS Number: 65-86-1
EINECS: 200-619-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR