4-Hydroxycoumarin

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C14230600
EHERC14230600EA 102 GBP
EHERC14230600
4-Hydroxycoumarin
4-Hydroxycoumarin
Formula: C₉H₆O₃
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 211…214 °C
MDL Number: MFCD00006856
CAS Number: 1076-38-6
EINECS: 214-060-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR