1-Heptadecanol

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Heptadecan-1-ol, Hexadecyl carbinol, 1-Heptadecanol

C14122700
EHERC14122700EA 116 GBP
EHERC14122700
1-Heptadecanol
1-Heptadecanol
Formula: H₃C(CH₂)₁₅CH₂OH
MW: 256.47 g/mol
Boiling Pt: 308.5 °C (1013 hPa)
Melting Pt: 53…57 °C
Density: 0.967 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002822
CAS Number: 1454-85-9
EINECS: 215-932-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR