n-Heptadecane

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

C14122500
EHERC14122500EA 89.7 GBP
EHERC14122500
n-Heptadecane
n-Heptadecane
Formula: H₃C(CH₂)₁₅CH₃
Boiling Pt: 302,1 °C (1013 hPa)
Melting Pt: 21,2 °C
Density: 0,7783 g/cm³ (20 °C)
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00009002
CAS Number: 629-78-7
EINECS: 211-108-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR