Eugenol

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

C13395000
EHERC13395000EA 102 GBP
EHERC13395000
Eugenol
Eugenol
Formula: C₁₀H₁₂O₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: ∼–10 °C
Density: 1.0652 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1
Merck Index: 13,03931

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR