2-Butoxyethanol

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether

C13328000
EHERC13328000EA 82.6 GBP
EHERC13328000
2-Butoxyethanol
2-Butoxyethanol
Formula: CH₃(CH₂)₃OCH₂CH₂OH
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00002884
CAS Number: 111-76-2
EINECS: 203-905-0
UN: 2810
ADR: 6.1,III
Merck Index: 13,01558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR