Dinoseb methyl ether

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C12811500
EHERC12811500EA 406 GBP
EHERC12811500
Dinoseb methyl ether
Dinoseb methyl ether
Formula: C₁₁H₁₄N₂O₅
MW: 254.24 g/mol
CAS Number: 6099-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR