α,α-Dichlorotoluene

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C12505000
EHERC12505000EA 121 GBP
EHERC12505000
α,α-Dichlorotoluene
α,α-Dichlorotoluene
Formula: (C₆H₅)CHCl₂
Boiling Pt: 205,2 °C (1013 hPa)
Melting Pt: –16,1 °C
Density: 1,2699 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000829
CAS Number: 98-87-3
EINECS: 202-709-2
UN: 1886
ADR: 6.1,II
Merck Index: 13,01056

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR