Diazinon

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

C12210000
EHERC12210000EA 142 GBP
EHERC12210000
Diazinon
Diazinon
Formula: C₁₂H₂₁N₂O₃PS
Boiling Pt: 306 °C (1013 hPa)
Density: 1.116 g/cm³ (20 °C)
Flash Pt: 104.4 °C (closed cup)
CAS Number: 333-41-5
EINECS: 206-373-8
UN: 3018
ADR: 6.1,III
Merck Index: 12,03043

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR