3,5-Dichloro-4-hydroxybenzonitrile

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C11550000
EHERC11550000EA 217 GBP
EHERC11550000
3,5-Dichloro-4-hydroxybenzonitrile
3,5-Dichloro-4-hydroxybenzonitrile
Formula: C₇H₃Cl₂NO
MDL Number: MFCD00002177
CAS Number: 1891-95-8
EINECS: 217-572-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR