2-Aminopyridine

Supplier: EHRENSTORFER
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C10220000
EHERC10220000EA 73.2 GBP
EHERC10220000
2-Aminopyridine
2-Aminopyridine
Formula: C₅H₆N₂
Boiling Pt: 210.6 °C (1013 hPa)
Melting Pt: 50…52 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006312
CAS Number: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1,II
Merck Index: 13,00472

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR