Silicone, high vacuum grease
Supplier: EDWARDS HIGH VACUUM METTLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H024-00-036
EDWAH024-00-036EA 67.6 GBP
EDWAH024-00-036
Silicone, high vacuum grease
Lubricants
A high vacuum grade grease, which is for use at system pressures lower than 10⁻⁶ mbar.