Scroll pump, dry, nXDS series
Supplier: EDWARDS HIGH VACUUM METTLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A73601983
EDWAA73601983EA 15310 GBP
EDWAA73601983
Scroll pump, dry, nXDS series
Pumps Vacuum Diaphragm Pumps
These nXDS series pumps are completely oil free, dry scroll and improved on their pumping speeds, combined with lower ultimate pressures, lower power consumption and lower noise.
  • Better work environment and low environmental impact
  • Advanced scroll-form and tip-seal technologies
  • Unique tip seal design
  • Simple operation and easy to use controls
  • Hermetically sealed for a lubricant-free vacuum environment
It also features the latest in tip seal technology giving significantly longer life between tip seal changes. Comes with integrated inverter drive withMore Product Information