Kits: amplification of DNA for fingerprinting
Supplier: EDVOTEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
130 130-C
EDVO130EA 99.1 GBP
EDVO130 EDVO130-C
Kits: amplification of DNA for fingerprinting
Educational Materials Forensics Educational Materials Forensic Kits
Ready-to-Load™ kit for teaching DNA fingerprinting and to show how DNA evidence is used in modern forensics. This experiment allows for varied results depending on the selection of DNA fingerprinting patterns.
  • For six lab groups
  • Complete in 50 minutes
Also required: Electrophoresis tank, power supply, automatic micropipette with tips, balance, microwave or hot plate, 65 °C water bath, white light visualisation system.
Delivery information: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable