Organic reference standard, ciprofloxacin, Ph.Eur., RS
Supplier: EDQM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Y0000198
EDQMY0000198EA 257 GBP
EDQMY0000198
Organic reference standard, ciprofloxacin, Ph.Eur., RS
Standards Organic Standards
  • In Monograph 1089
  • Soluble in/miscibility with hot DMSO and hot methanol