Robust Temperature Bottle Data Loggers, EBI 12-T46X
Supplier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1340-6623 1340-6624 1340-6625 1340-6626 1340-6627
EBRO1340-6623EA 3670 GBP
EBRO1340-6623 EBRO1340-6624 EBRO1340-6625 EBRO1340-6626 EBRO1340-6627
Robust Temperature Bottle Data Loggers, EBI 12-T46X
Data Loggers
Designed for monitoring both the interior and exterior of bottles or cans, this data logger is essential for validating manufacturing processes. The central axial probe accurately measures the internal temperature, while the lateral sensor captures the external temperature of the packaging. The collected data allows for precise analysis of temperature exposure duration. Known for its durability and high accuracy, this logger is a reliable tool in quality assurance.
  • Wide measuring range: −55 to +140 °C
  • High-quality PT1000 sensor (class A)
  • Rigid, external
More Product Information