High Temperature Data Loggers, EBI 12-T230
Supplier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1340-6606 1340-6607 1340-6608 1340-6609
EBRO1340-6606EA 3240 GBP
EBRO1340-6606 EBRO1340-6607 EBRO1340-6608 EBRO1340-6609
High Temperature Data Loggers, EBI 12-T230
Data Loggers
The EBI 12-T230 is a robust temperature data logger designed for food monitoring, particularly during pasteurisation processes. With a measurement range from –55 to +150 °C and an accuracy of up to ±0,1 °C, it ensures reliable and precise temperature readings in demanding applications.
  • Suitable for steam sterilisers, washer-disinfectors
  • Tempreture probe, axial pointed with M5 thread
Its durable stainless steel housing make it a reliable solution for ensuring temperature control in critical food processing tasks. With a memory capacity of 27,840More Product Information