Safety underbench cabinets, type 90, UTS ergo line MT-5, 500 mm depth
Supplier: DUPERTHAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
29-060959-001 29-060957-401 29-060957-001
DUEH29-060959-001EA 2050 GBP
DUEH29-060959-001 DUEH29-060957-401 DUEH29-060957-001
Safety underbench cabinets, type 90, UTS ergo line MT-5, 500 mm depth
Furniture Storage Cabinets
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position andMore Product Information