Safety underbench cabinets, type 90, UTS ergo line S, 600 mm depth
Supplier: DUPERTHAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
29-060669-503 29-060669-403 29-060669-003 29-060667-403 29-060667-003 29-060667-503
DUEH29-060669-503EA 1500 GBP
DUEH29-060669-503 DUEH29-060669-403 DUEH29-060669-003 135-1896 DUEH29-060667-003 DUEH29-060667-503
Safety underbench cabinets, type 90, UTS ergo line S, 600 mm depth
Furniture Storage Cabinets
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position andMore Product Information